siRNA Details
| Gene Symbol: Ndufb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033305.2 | NP_001028477.1 | 2, 3 | 312 | ||
| GenBank mRNA | AI853615.1 | - | 2, 3 | 236 | ||
| AK146762.1 | - | 2, 3 | 314 | |||
| BC147062.1 | - | 2, 3 | 301 | 9056053 | ||
| BC147063.1 | - | 2, 3 | 301 | 9056059 | ||

Ndufb6
Genomic Map