siRNA Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172381.2 | NP_759013.2 | 49 | 5646 | ||
| GenBank mRNA | AK030369.1 | - | 9 | 1075 | ||
| AK129127.1 | - | 32 | 3586 | |||
| AK208848.1 | - | 4 | 248 | |||
| BC025035.1 | - | 19 | 1918 | |||
| BC058684.1 | - | 49 | 5646 | 6834912 | ||

AI314180
Genomic Map