siRNA Details
| Gene Symbol: Ttc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177667.4 | NP_808335.1 | 1 | 564 | ||
| GenBank mRNA | AK028895.1 | - | 1 | 565 | ||
| BC132162.1 | - | 1 | 505 | 40130439 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc22| Gene Symbol: Ttc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177667.4 | NP_808335.1 | 1 | 564 | ||
| GenBank mRNA | AK028895.1 | - | 1 | 565 | ||
| BC132162.1 | - | 1 | 505 | 40130439 | ||