siRNA Details
| Gene Symbol: Tesk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146151.4 | NP_666263.3 | 9 | 1249 | ||
| GenBank mRNA | AK031814.1 | - | 9 | 1242 | ||
| AK047755.1 | - | 9 | 1250 | |||
| AK134575.1 | - | 5 | 355 | |||
| BC019149.1 | - | 8 | 982 | 5053345 | ||
| BC029766.1 | - | 9 | 1227 | 4947564 | ||

Tesk2
Genomic Map