siRNA Details
| Gene Symbol: Szt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198170.4 | NP_937813.3 | 15 | 2192 | ||
| GenBank mRNA | FJ998170.1 | - | 15 | 2192 | ||
| GU433214.1 | - | 15 | 2118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Szt2| Gene Symbol: Szt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198170.4 | NP_937813.3 | 15 | 2192 | ||
| GenBank mRNA | FJ998170.1 | - | 15 | 2192 | ||
| GU433214.1 | - | 15 | 2118 | |||