siRNA Details
Gene Symbol: Szt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198170.4 | NP_937813.3 | 29 | 4256 | ||
GenBank mRNA | FJ998170.1 | - | 29 | 4256 | ||
GU433214.1 | - | 29 | 4182 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Szt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198170.4 | NP_937813.3 | 29 | 4256 | ||
GenBank mRNA | FJ998170.1 | - | 29 | 4256 | ||
GU433214.1 | - | 29 | 4182 |