siRNA Details
Gene Symbol: Tmem125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172383.3 | NP_759015.2 | 4 | 838 | ||
GenBank mRNA | AK050146.1 | - | 4 | 842 | ||
AK160304.1 | - | 2 | 493 | |||
AK162829.1 | - | 453 | ||||
AK162907.1 | - | 2 | 509 | |||
BC039806.1 | - | 1 | 336 | 1516024 |
