siRNA Details
| Gene Symbol: Rhbdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183163.2 | NP_898986.2 | 2 | 147 | ||
| GenBank mRNA | AK078969.1 | - | 2 | 149 | ||
| BB625403.1 | - | 2 | 147 | |||
| BC115494.1 | - | 2 | 119 | |||
| BC115495.1 | - | 2 | 119 | |||
| EH099096.1 | - | 2 | 140 | |||

Rhbdl2
Genomic Map