siRNA Details
| Gene Symbol: Eva1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172145.3 | NP_742157.1 | 3 | 741 | ||
| GenBank mRNA | AK076006.1 | - | 3 | 743 | ||
| AK077984.1 | - | 3 | 1069 | |||
| AK186422.1 | - | 224 | ||||
| BC029204.1 | - | 2 | 374 | 3964022 | ||
More Information
Other Species Targeted
Set Membership: |
|

Eva1b
Genomic Map