siRNA Details
| Gene Symbol: Adgrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173071.3 | NP_775094.2 | 22, 23 | 3582 | ||
| GenBank mRNA | BC056926.1 | - | 22, 23 | 3496 | 5705854 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adgrb2| Gene Symbol: Adgrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173071.3 | NP_775094.2 | 22, 23 | 3582 | ||
| GenBank mRNA | BC056926.1 | - | 22, 23 | 3496 | 5705854 | |
Set Membership: |
|