siRNA Details
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198305.2 | NP_938047.1 | 3 | 537 | ||
| GenBank mRNA | AK049214.1 | - | 3 | 481 | ||
| AK154523.1 | - | 3 | 475 | |||
| AK170825.1 | - | 3 | 475 | |||
| AY423764.1 | - | 3 | 515 | |||
| BC139309.1 | - | 3 | 478 | 8862331 | ||

Klhl17
Genomic Map