siRNA Details
| Gene Symbol: 4931409K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177676.6 | NP_808344.3 | 16 | 1973 | ||
| GenBank mRNA | BC137999.1 | - | 12 | 1694 | 8861018 | |
| BC145792.1 | - | 12 | 1696 | 40131095 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931409K22Rik| Gene Symbol: 4931409K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177676.6 | NP_808344.3 | 16 | 1973 | ||
| GenBank mRNA | BC137999.1 | - | 12 | 1694 | 8861018 | |
| BC145792.1 | - | 12 | 1696 | 40131095 | ||