siRNA Details
| Gene Symbol: 4931409K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177676.6 | NP_808344.3 | 18 | 2313 | ||
| GenBank mRNA | AK029839.1 | - | 17 | 2745 | ||
| AV271477.2 | - | 2 | 259 | |||
| BC137999.1 | - | 14 | 2034 | 8861018 | ||
| BC145792.1 | - | 14 | 2036 | 40131095 | ||

4931409K22Rik
Genomic Map