siRNA Details
| Gene Symbol: Kmt2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081383.1 | NP_001074852.1 | 52 | 13192 | ||
| GenBank mRNA | AK129377.1 | - | 11 | 2819 | ||
| AK136029.1 | - | 5 | 843 | |||
| AY036887.1 | - | 2 | 317 | |||
| AY138582.1 | - | 47 | 8807 | |||

Kmt2c
Genomic Map