siRNA Details
| Gene Symbol: Kmt2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081383.1 | NP_001074852.1 | 55 | 14508 | ||
| GenBank mRNA | AK129377.1 | - | 14 | 4123 | ||
| AK136029.1 | - | 8 | 2159 | |||
| AK148595.1 | - | 4 | 515 | |||
| AK161051.1 | - | 4 | 1253 | |||
| AK164521.1 | - | 4 | 772 | |||
| AY036887.1 | - | 5 | 1621 | |||
| AY138582.1 | - | 50 | 10111 | |||

Kmt2c
Genomic Map