siRNA Details
| Gene Symbol: Dnajc5g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177677.3 | NP_808345.1 | 5 | 589 | ||
| GenBank mRNA | AK076638.1 | - | 5 | 590 | ||
| BC089502.1 | - | 5 | 568 | 6774833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc5g| Gene Symbol: Dnajc5g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177677.3 | NP_808345.1 | 5 | 589 | ||
| GenBank mRNA | AK076638.1 | - | 5 | 590 | ||
| BC089502.1 | - | 5 | 568 | 6774833 | ||