siRNA Details
| Gene Symbol: Atp10d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153389.3 | NP_700438.3 | 23 | 4249 | ||
| GenBank mRNA | AJ441079.1 | - | 23 | 4249 | ||
| AK036646.1 | - | 22 | 3911 | |||
| AK133052.1 | - | 12 | 1700 | |||
| AK144558.1 | - | 5 | 634 | |||
| AK162523.1 | - | 2 | 236 | |||
| BC029551.1 | - | 6 | 708 | |||
| BC064739.1 | - | 7 | 801 | |||
| BC141328.1 | - | 23 | 4100 | 9055940 | ||

Atp10d
Genomic Map