siRNA Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145561.2 | NP_663536.1 | 2 | 137 | ||
| GenBank mRNA | AB053953.1 | - | 2 | 171 | ||
| AF448809.1 | - | 2 | 116 | |||
| BC020151.1 | - | 2 | 137 | 4021767 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11d| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145561.2 | NP_663536.1 | 2 | 137 | ||
| GenBank mRNA | AB053953.1 | - | 2 | 171 | ||
| AF448809.1 | - | 2 | 116 | |||
| BC020151.1 | - | 2 | 137 | 4021767 | ||