siRNA Details
| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177680.3 | NP_808348.2 | 8 | 1262 | ||
| GenBank mRNA | AK173312.1 | - | 5 | 525 | ||
| BC022697.1 | - | 1 | 55 | |||
| BC037593.1 | - | 2 | 155 | |||
| BC044650.1 | - | 6 | 1080 | |||
| BC048817.1 | - | 1 | 40 | |||
| CN526522.1 | - | 5 | 586 | |||
| CO424360.1 | - | 3 | 207 | |||

Ythdc1
Genomic Map