siRNA Details
| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177680.3 | NP_808348.2 | 9, 10 | 1434 | ||
| GenBank mRNA | AB221619.1 | - | 1, 2 | 58 | ||
| AI121974.1 | - | 2, 3 | 169 | |||
| AK173312.1 | - | 6, 7 | 697 | |||
| BC022697.1 | - | 2, 3 | 227 | |||
| BC037593.1 | - | 3, 4 | 327 | |||
| BC044650.1 | - | 7, 8 | 1252 | |||
| BC048817.1 | - | 2, 3 | 212 | |||
| CO424360.1 | - | 4, 5 | 379 | |||

Ythdc1
Genomic Map