siRNA Details
| Gene Symbol: Cox18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033310.3 | NP_001028482.2 | 5 | 856 | ||
| NM_001163456.1 | NP_001156928.1 | 5 | 859 | |||
| GenBank mRNA | AK156053.1 | - | 5 | 825 | ||
| BC021612.1 | - | 3 | 407 | |||
| BC038311.1 | - | 5 | 629 | |||
| BC138972.1 | - | 5 | 834 | 8861994 | ||
| BC145479.1 | - | 5 | 837 | 9054042 | ||
| BC145486.1 | - | 5 | 838 | |||
| BC145728.1 | - | 5 | 834 | 40131031 | ||
| DV661727.1 | - | 4 | 467 | |||

Cox18
Genomic Map