siRNA Details
| Gene Symbol: Agfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155221.1 | - | 4 | 670 | ||
| AK171048.1 | - | 4 | 715 | |||
| BC003330.1 | - | 4 | 682 | 3256586 | ||
| BC046788.1 | - | 4 | 606 | 5716604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agfg2| Gene Symbol: Agfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155221.1 | - | 4 | 670 | ||
| AK171048.1 | - | 4 | 715 | |||
| BC003330.1 | - | 4 | 682 | 3256586 | ||
| BC046788.1 | - | 4 | 606 | 5716604 | ||