siRNA Details
| Gene Symbol: BC037034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153161.3 | NP_694801.2 | 10 | 1759 | ||
| GenBank mRNA | AK046882.1 | - | 6 | 2782 | ||
| AK138960.1 | - | 10 | 1703 | |||
| AK213494.1 | - | 1 | 25 | |||
| AK216902.1 | - | 1 | 78 | |||
| BC037034.1 | - | 9 | 1162 | 4505093 | ||
| BC040828.1 | - | 1 | 1613 | |||
| BC044860.1 | - | 1 | 1625 | |||

BC037034
Genomic Map