siRNA Details
| Gene Symbol: Uspl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013378.2 | NP_001013396.2 | 8 | 1565 | ||
| NM_001115149.1 | NP_001108621.1 | 8 | 1565 | |||
| NM_001115150.1 | NP_001108622.1 | 7 | 1396 | |||
| NM_001115151.1 | NP_001108623.1 | 7 | 1396 | |||
| NM_001115153.1 | NP_001108625.1 | 9 | 1742 | |||
| NM_001286824.1 | NP_001273753.1 | 8 | 1523 | |||
| NM_001286825.1 | NP_001273754.1 | 8 | 1507 | |||
| GenBank mRNA | AK050969.1 | - | 8 | 1549 | ||
| AK088787.1 | - | 7 | 1262 | |||
| AK145409.1 | - | 9 | 1640 | |||
| AK146820.1 | - | 8 | 1390 | |||
| AK161654.1 | - | 7 | 1243 | |||
| AK168702.1 | - | 8 | 1390 | |||
| AK168981.1 | - | 8 | 1417 | |||
| BC028849.1 | - | 8 | 1459 | |||
| BC032291.1 | - | 5 | 3535 | |||
| BC086664.1 | - | 8 | 1407 | |||

Uspl1
Genomic Map