siRNA Details
| Gene Symbol: Fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153573.1 | NP_705801.1 | 3, 4 | 553 | ||
| GenBank mRNA | AK030620.1 | - | 3, 4 | 604 | ||
| AK081692.1 | - | 3, 4 | 580 | |||
| AK163384.1 | - | 699 | ||||
| BC024846.1 | - | 980 | ||||
| BC029109.1 | - | 3, 4 | 553 | 3985764 | ||
| BC034313.1 | - | 4, 5 | 668 | 5365046 | ||

Fkbp14
Genomic Map