siRNA Details
| Gene Symbol: Ccser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164316.1 | NP_001157788.1 | 2 | 822 | ||
| NM_183310.2 | NP_899133.2 | 2 | 874 | |||
| GenBank mRNA | AK031754.1 | - | 2 | 610 | ||
| BC107398.2 | - | 2 | 661 | 40058350 | ||
| BC107399.1 | - | 2 | 660 | 40058352 | ||
| BC117738.2 | - | 2 | 661 | 40089965 | ||

Ccser1
Genomic Map