siRNA Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033929.2 | NP_001029101.1 | 4 | 648 | ||
| NM_178413.5 | NP_848500.3 | 5 | 716 | |||
| GenBank mRNA | AK165463.1 | - | 4 | 614 | ||
| BC025604.1 | - | 5 | 679 | 5250319 | ||
| BC051244.1 | - | 4 | 610 | 4975661 | ||

Thnsl2
Genomic Map