siRNA Details
| Gene Symbol: Mob1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145571.2 | NP_663546.1 | 2 | 325 | ||
| GenBank mRNA | AK028416.1 | - | 2 | 348 | ||
| AK028695.1 | - | 2 | 286 | |||
| AK158228.1 | - | 2 | 308 | |||
| AK163300.1 | - | 2 | 347 | |||
| AK167515.1 | - | 2 | 320 | |||
| AK170691.1 | - | 2 | 319 | |||
| AK172205.1 | - | 2 | 328 | |||
| AK213414.1 | - | 1 | 89 | |||
| AK216221.1 | - | 1 | 16 | |||
| BC009149.1 | - | 2 | 325 | 2651599 | ||
| BC033463.1 | - | 2 | 279 | 3500664 | ||

Mob1a
Genomic Map