siRNA Details
| Gene Symbol: Mob1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145571.2 | NP_663546.1 | 2 | 381 | ||
| GenBank mRNA | AK028416.1 | - | 2 | 404 | ||
| AK028695.1 | - | 2 | 342 | |||
| AK158228.1 | - | 2 | 364 | |||
| AK163300.1 | - | 2 | 403 | |||
| AK167515.1 | - | 2 | 376 | |||
| AK170691.1 | - | 2 | 375 | |||
| AK172205.1 | - | 2 | 384 | |||
| AK216221.1 | - | 1 | 72 | |||
| BC009149.1 | - | 2 | 381 | 2651599 | ||
| BC033463.1 | - | 2 | 335 | 3500664 | ||

Mob1a
Genomic Map