siRNA Details
| Gene Symbol: Mob1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145571.2 | NP_663546.1 | 4 | 508 | ||
| GenBank mRNA | AK028416.1 | - | 4 | 531 | ||
| AK158228.1 | - | 4 | 491 | |||
| AK167515.1 | - | 4 | 503 | |||
| AK170691.1 | - | 4 | 502 | |||
| AK172205.1 | - | 4 | 511 | |||
| AK216221.1 | - | 3 | 199 | |||
| BC009149.1 | - | 4 | 508 | 2651599 | ||
| BC033463.1 | - | 4 | 462 | 3500664 | ||

Mob1a
Genomic Map