siRNA Details
Gene Symbol: Zfp637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177684.2 | NP_808352.1 | 2 | 126 | ||
GenBank mRNA | AB221671.1 | - | 1 | 29 | ||
AK134044.1 | - | 2 | 166 | |||
AK146789.1 | - | 2 | 217 | |||
BC037994.1 | - | 1 | 105 | |||
BC048843.1 | - | 2 | 126 | 5031082 |
