siRNA Details
| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177687.3 | NP_808355.1 | 3 | 572 | ||
| GenBank mRNA | AK038845.1 | - | 1 | 50 | ||
| AK045467.1 | - | 3 | 573 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crebl2| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177687.3 | NP_808355.1 | 3 | 572 | ||
| GenBank mRNA | AK038845.1 | - | 1 | 50 | ||
| AK045467.1 | - | 3 | 573 | |||