siRNA Details
| Gene Symbol: Pyroxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183165.3 | NP_898988.2 | 11 | 1212 | ||
| GenBank mRNA | AK148411.1 | - | 1251 | |||
| AK165677.1 | - | 11 | 1215 | |||
| BC022914.1 | - | 9 | 2642 | |||
| BC027061.1 | - | 11 | 1237 | 5355217 | ||

Pyroxd1
Genomic Map