siRNA Details
| Gene Symbol: Gys2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145572.2 | NP_663547.2 | 16 | 2339 | ||
| GenBank mRNA | AK138992.1 | - | 16 | 2047 | ||
| AK149391.1 | - | 16 | 2064 | |||
| BC021322.1 | - | 16 | 2200 | 5051685 | ||
| BC158081.1 | - | 16 | 2200 | 9007373 | ||
| BX516187.1 | - | 1 | 41 | |||

Gys2
Genomic Map