siRNA Details
| Gene Symbol: Klhl42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081237.1 | NP_001074706.1 | 3 | 1824 | ||
| GenBank mRNA | AK048908.1 | - | 3 | 1413 | ||
| AK052765.1 | - | 3 | 1401 | |||
| AK083047.1 | - | 3 | 1409 | |||
| AK129335.1 | - | 3 | 734 | |||
| BC037084.1 | - | 3 | 1429 | |||
| BC147633.1 | - | 3 | 1580 | 9056623 | ||
| BC147636.1 | - | 3 | 1579 | 9056626 | ||
| BC151064.1 | - | 3 | 1580 | 9087977 | ||
| BC151065.1 | - | 3 | 1580 | 9087978 | ||
| BC151068.1 | - | 3 | 1580 | 9087981 | ||
| BC151076.1 | - | 3 | 1580 | 9087989 | ||

Klhl42
Genomic Map