siRNA Details
| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201378.1 | NP_001188307.1 | 13 | 2567 | ||
| NM_145574.3 | NP_663549.3 | 13 | 2567 | |||
| GenBank mRNA | AK160245.1 | - | 6 | 1294 | ||
| BC006583.1 | - | 11 | 2155 | |||
| EU029664.1 | - | 13 | 2567 | |||
| EU029665.1 | - | 13 | 2567 | |||

Ccdc136
Genomic Map