siRNA Details
| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145574.3 | NP_663549.3 | 14, 15 | 2844 | ||
| GenBank mRNA | AK160245.1 | - | 7, 8 | 1571 | ||
| EU029664.1 | - | 14, 15 | 2844 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc136| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145574.3 | NP_663549.3 | 14, 15 | 2844 | ||
| GenBank mRNA | AK160245.1 | - | 7, 8 | 1571 | ||
| EU029664.1 | - | 14, 15 | 2844 | |||