siRNA Details
| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201378.1 | NP_001188307.1 | 4 | 712 | ||
| NM_145574.3 | NP_663549.3 | 4 | 712 | |||
| GenBank mRNA | BC006583.1 | - | 2 | 300 | ||
| CX222432.1 | - | 1 | 272 | |||
| CX564275.1 | - | 1 | 217 | |||
| EU029664.1 | - | 4 | 712 | |||
| EU029665.1 | - | 4 | 712 | |||

Ccdc136
Genomic Map