siRNA Details
| Gene Symbol: Shisa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290291.1 | NP_001277220.1 | 5 | 1615 | ||
| NM_172737.4 | NP_766325.3 | 6 | 1666 | |||
| GenBank mRNA | AK085127.1 | - | 5 | 888 | ||
| AK145016.1 | - | 4 | 900 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shisa7| Gene Symbol: Shisa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290291.1 | NP_001277220.1 | 5 | 1615 | ||
| NM_172737.4 | NP_766325.3 | 6 | 1666 | |||
| GenBank mRNA | AK085127.1 | - | 5 | 888 | ||
| AK145016.1 | - | 4 | 900 | |||