siRNA Details
| Gene Symbol: Zfp628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170759.2 | NP_739565.2 | 3 | 1838 | ||
| GenBank mRNA | AF435832.1 | - | 1 | 1542 | ||
| AK048291.1 | - | 3 | 1805 | |||
| AK155214.1 | - | 3 | 1838 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp628| Gene Symbol: Zfp628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170759.2 | NP_739565.2 | 3 | 1838 | ||
| GenBank mRNA | AF435832.1 | - | 1 | 1542 | ||
| AK048291.1 | - | 3 | 1805 | |||
| AK155214.1 | - | 3 | 1838 | |||