siRNA Details
| Gene Symbol: Ccdc106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290429.1 | NP_001277358.1 | 5, 6 | 742 | ||
| NM_001290432.1 | NP_001277361.1 | 5, 6 | 794 | |||
| NM_146178.2 | NP_666290.2 | 4, 5 | 1426 | |||
| GenBank mRNA | AK145417.1 | - | 5, 6 | 743 | ||
| BC018462.1 | - | 4, 5 | 956 | 3501538 | ||
| BC036277.1 | - | 4, 5 | 956 | |||

Ccdc106
Genomic Map