siRNA Details
| Gene Symbol: Ccdc106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290429.1 | NP_001277358.1 | 6 | 873 | ||
| NM_001290432.1 | NP_001277361.1 | 6 | 925 | |||
| NM_146178.2 | NP_666290.2 | 5 | 1557 | |||
| GenBank mRNA | AB041804.1 | - | 4 | 2046 | ||
| AK145417.1 | - | 6 | 874 | |||
| AK148389.1 | - | 1712 | ||||
| BC018462.1 | - | 5 | 1087 | 3501538 | ||
| BC036277.1 | - | 5 | 1087 | |||

Ccdc106
Genomic Map