siRNA Details
| Gene Symbol: Ccdc106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290429.1 | NP_001277358.1 | 2, 3 | 240 | ||
| NM_001290432.1 | NP_001277361.1 | 2, 3 | 292 | |||
| NM_146178.2 | NP_666290.2 | 1, 2 | 924 | |||
| GenBank mRNA | AK145417.1 | - | 2, 3 | 241 | ||
| BC018462.1 | - | 1, 2 | 454 | 3501538 | ||
| BC036277.1 | - | 1, 2 | 454 | |||
| BY267627.1 | - | 2, 3 | 366 | |||
| CX732283.1 | - | 2, 3 | 293 | |||

Ccdc106
Genomic Map