siRNA Details
| Gene Symbol: Zfp114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029933.2 | NP_001025104.2 | 4 | 890 | ||
| GenBank mRNA | AF167317.1 | - | 4 | 875 | ||
| AK139475.1 | - | 3 | 1809 | |||
| AY166793.1 | - | 2 | 652 | |||
| BC096771.1 | - | 2 | 1116 | |||
| BC138311.1 | - | 2 | 1005 | 8861331 | ||
| BC138312.1 | - | 2 | 1005 | 8861332 | ||

Zfp114
Genomic Map