siRNA Details
| Gene Symbol: Cxcl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153576.2 | NP_705804.2 | 3 | 334 | ||
| GenBank mRNA | AK143005.1 | - | 3 | 250 | ||
| AY374329.1 | - | 3 | 237 | |||
| AY598463.1 | - | 3 | 335 | |||
| BC024561.1 | - | 3 | 334 | 5004999 | ||

Cxcl17
Genomic Map