siRNA Details
| Gene Symbol: Zfp420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172740.2 | NP_766328.2 | 2 | 151 | ||
| GenBank mRNA | AK029387.1 | - | 2 | 127 | ||
| AK045821.1 | - | 2 | 64 | |||
| BC055817.1 | - | 2 | 71 | 3595825 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp420| Gene Symbol: Zfp420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172740.2 | NP_766328.2 | 2 | 151 | ||
| GenBank mRNA | AK029387.1 | - | 2 | 127 | ||
| AK045821.1 | - | 2 | 64 | |||
| BC055817.1 | - | 2 | 71 | 3595825 | ||