siRNA Details
| Gene Symbol: Zfp420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172740.2 | NP_766328.2 | 4 | 227 | ||
| GenBank mRNA | AK045821.1 | - | 4 | 140 | ||
| BC050249.1 | - | 3 | 112 | |||
| BC055817.1 | - | 4 | 147 | 3595825 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp420| Gene Symbol: Zfp420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172740.2 | NP_766328.2 | 4 | 227 | ||
| GenBank mRNA | AK045821.1 | - | 4 | 140 | ||
| BC050249.1 | - | 3 | 112 | |||
| BC055817.1 | - | 4 | 147 | 3595825 | ||