siRNA Details
| Gene Symbol: Syne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290565.1 | NP_001277494.1 | 5 | 786 | ||
| NM_153577.2 | NP_705805.1 | 6 | 1035 | |||
| GenBank mRNA | AK002612.1 | - | 1408 | |||
| BC004761.1 | - | 6 | 984 | 3155758 | ||
| BC023803.1 | - | 6 | 1022 | 5352953 | ||
| BC023884.1 | - | 1 | 596 | |||
| BC056649.1 | - | 6 | 974 | 5036575 | ||
| BG063039.2 | - | 3 | 316 | |||

Syne4
Genomic Map