siRNA Details
| Gene Symbol: Zfp939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243021.1 | NP_001229950.1 | 5 | 1164 | ||
| GenBank mRNA | AK034698.1 | - | 5 | 1162 | ||
| AK162098.1 | - | 1 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp939| Gene Symbol: Zfp939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243021.1 | NP_001229950.1 | 5 | 1164 | ||
| GenBank mRNA | AK034698.1 | - | 5 | 1162 | ||
| AK162098.1 | - | 1 | 194 | |||