siRNA Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271879.1 | NP_001258808.1 | 12 | 1377 | ||
| NM_177694.6 | NP_808362.2 | 13 | 1419 | |||
| GenBank mRNA | AB125740.1 | - | 13 | 1419 | ||
| AB206762.1 | - | 13 | 1419 | |||
| AK034197.1 | - | 13 | 1401 | |||
| BC109163.2 | - | 12 | 1299 | 40047987 | ||

Ano5
Genomic Map